missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ OIML Class F1 Reference Line Wire Weight with DAkkS Certificate

Constructed from specialty stainless steel. Sartorius™ OIML Class F1 Wire Weight with DAkkS Certificate is non-magnetizable with a density of 7,950 kg/m3.

Brand:  Sartorius™ YCW0131-02

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15346055

  • £50.20 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Polished, special stainless steel construction with density of 7,950kg/m3
  • Highly corrosion resistant
  • Non-magnetizable
  • Tolerances comply with OIML R111-1
  • DAkkS Certified
Specifications

Specifications

Reference Line Milligram Weight
OIML Class F1
7,950kg/m3
DAkkS calibration certificate
Aluminum
1mg
Videos