missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ OIML Class F1 Proof Line Weight Set without Certificate
Includes the same features and properties as the individual weights of this series, along with the corresponding maximum permissible errors. Sartorius™ OIML Class F1 Proof LIne Weight Set without Certificate is supplied with gloves, tweezers and brush in high-quality plastic case
£281.00 - £2530.00
Product Code | Brand | Mass | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Material | Price | Quantity & Availability | ||||
11363354
|
Sartorius™
YCS01-253-AC-00 |
1 mg to 500 mg | Aluminum (1 to 5 mg), Nickel Silver (10 to 500 mg) |
£281.00
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
11333354
|
Sartorius™
YCS31-513-AC-00 |
1 g to 100 g | Special Stainless Steel |
£575.00
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
11323374
|
Sartorius™
YCS31-613-AC-00 |
1 g to 1 kg | Special Stainless Steel |
£1240.00
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
11303384
|
Sartorius™
YCS31-653-AC-00 |
1 g to 5 kg | Special Stainless Steel |
£2530.00
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
Description
Values 1 to 5mg: Aluminum leaf weights with density of 2,650 kg/m3Values of 10 to 500mg: Nickel silver construction with density of 8,600 kg/m3.
Values of 1g and higher: Polished, special stainless steel knob weights with density of 7,950kg/m3
- Highly corrosion resistant
- Non-magnetizable
- Tolerances comply with OIML R111-1
- Includes sturdy plastic carry case
Specifications
Proof Line Weight Set | |
OIML Class F1 |