missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ OIML Class F1 Proof Line Knob Weight with DAkkS Certificate
Polished non-magnetizable calibration weights. Sartorius™ OIML Class F1 Proof Line Knob Weight with DAkkS Certificate is constructed of special steel and supplied in a plastic case.
£118.00 - £1090.00
Specifications
Product Type | Proof Line Weight |
---|---|
Certifications/Compliance | DAkkS calibration certificate |
Classification | OIML Class F1 |
Material | Special Stainless Steel |
Density | 7,950kg/m3 |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
11323324
|
Sartorius™
YCW313-AC-02 |
1 g |
£121.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11333324
|
Sartorius™
YCW323-AC-02 |
2 g |
£118.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11343324
|
Sartorius™
YCW353-AC-02 |
5 g |
£121.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11353324
|
Sartorius™
YCW413-AC-02 |
10 g |
£126.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11363324
|
Sartorius™
YCW423-AC-02 |
20 g |
£135.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11373324
|
Sartorius™
YCW453-AC-02 |
50 g |
£149.00
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
11383324
|
Sartorius™
YCW513-AC-02 |
100 g |
£165.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11393324
|
Sartorius™
YCW523-AC-02 |
200 g |
£161.00
Each |
In Stock
Log in to see stock. |
|||||
11303334
|
Sartorius™
YCW553-AC-02 |
500 g |
£286.00
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
11313334
|
Sartorius™
YCW613-AC-02 |
1 kg |
£310.00
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
11323334
|
Sartorius™
YCW623-AC-02 |
2 kg |
£455.00
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||||
11333334
|
Sartorius™
YCW653-AC-02 |
5 kg |
£630.00
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
11377494
|
Sartorius™
YCW713-AC-02 |
10 kg |
£1090.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- Non-magnetizable
- Supplied in a high-quality plastic case (unless otherwise specified) suitable for use in cleanrooms
- Special Stainless steel construction with a density of 7,950 kg/m3
- Complies with OIML R111-1 International Recommentation
- DAkkS Certified
Specifications
Proof Line Weight | |
OIML Class F1 | |
7,950kg/m3 |
DAkkS calibration certificate | |
Special Stainless Steel |