missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Adventurer™ Pro Balances

Industrial, laboratory-grade balances for the classroom.

Brand:  OHAUS™ 80103580

UNSPSC : 41110000

Code : A0

Additional Details : Weight : 2.28000kg

 View more versions of this product

Product Code. 10452772

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications Description & Specifications

Specifications

Precision Balance
ISO 9001, GLP, GMP, CE, FCC, cULus, CCSAUS, NRTL
25.4cm
7.2cm
Automatic
0.1g
ABS Plastic, Stainless Steel, Glass
10 to 30°C
19.3cm
Carat, grain, pound, ounce, pennyweight, gram, troy ounce, kilogram
4100g
RS232, USB
4 Button keypad with dedicated yes, no, back and exit functions
0.1g
AC adapter, RS232 interface, in-use cover, integral security bracket, weigh below hook, manual
±0.2g
AV4101
2 sec.
1.6kg
100/120V 50/60Hz
2 Years
LCD
Alternative Products