Learn More
Tocris Bioscience™ OGT 2115
Antiangiogenic. Heparanase inhibitor
Brand:  Tocris Bioscience™ 27101
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13441784
  • £91.14 / 1mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Heparanase inhibitor (IC50 = 0.4 μM) that displays no major inhibition of human cytochrome P450 isoenzymes (IC50 > 30 μM). Exhibits antiangiogenic properties in vitro (IC50 = 1 μM).
Specifications
Specifications
2-[4-[[3-(4-Bromophenyl)-1-oxo-2-propenyl]amino]-3-fluorophenyl]-5-benzoxazoleacetic acid
853929-59-6
C24H16BrFN2O4
1mg
0.97
>98%
Observed MW: 495.3
Soluble to 10mM in DMSO