missing translation for 'onlineSavingsMsg'
Learn More

ODQ, Tocris Bioscience™

Selective inhibitor of NO-sensitive guanylyl cyclase

Brand:  Tocris (Bio-Techne) 0880/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13438103

  • £410.13 / 50mg

Estimated Shipment date
from Supplier 23-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A potent and selective inhibitor of NO-sensitive guanylyl cyclase.
Specifications

Chemical Identifiers

41443-28-1
187.158
1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq
[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
C9H5N3O2
LZMHWZHOZLVYDL-UHFFFAOYSA-N
1456
C1=CC=C2C(=C1)N=CC3=NOC(=O)N23

Specifications

50mg
41443-28-1
1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq
C1=CC=C2C(=C1)N=CC3=NOC(=O)N23
187.158
187.16
ODQ
C9H5N3O2
LZMHWZHOZLVYDL-UHFFFAOYSA-N
[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
1456
>99%
Videos
Documents