missing translation for 'onlineSavingsMsg'
Learn More

Octylmethyldichlorosilane, 97%, Thermo Scientific™

Catalog Number p-177071
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
14799-93-0
C9H20Cl2Si
227.24
MFCD00042606
QHBMMABVNRSRHW-UHFFFAOYSA-N
n-octylmethyldichlorosilane, silane, dichloromethyloctyl, dichloro methyl-n-octylsilane, methyloctyldichlorosilane, dichloro methyl octyl silane, dichloro methyl octylsilane, octylmethyldichlorosilane, acmc-209czn, methyloctyl dichlorosilane, dichloro methyl octylsilane #
84649
dichloro-methyl-octylsilane
CCCCCCCC[Si](C)(Cl)Cl
This item is not returnable. View return policy
14799-93-0
C9H20Cl2Si
227.24
MFCD00042606
QHBMMABVNRSRHW-UHFFFAOYSA-N
n-octylmethyldichlorosilane, silane, dichloromethyloctyl, dichloro methyl-n-octylsilane, methyloctyldichlorosilane, dichloro methyl octyl silane, dichloro methyl octylsilane, octylmethyldichlorosilane, acmc-209czn, methyloctyl dichlorosilane, dichloro methyl octylsilane #
84649
dichloro-methyl-octylsilane
CCCCCCCC[Si](C)(Cl)Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 230°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C8H17Si(CH3)Cl2
Refractive Index 1.442
Quantity 5 mL
Specific Gravity 0.97
Solubility Information Solubility in water: .. Other solubilities: soluble in non-protic solvents,e.g. aliphatic,or aromatic(chlorinated) hydrocarbons
Formula Weight 227.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Octylmethyldichlorosilane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Octylmethyldichlorosilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'