missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octylmagnesium bromide, 2M solution in diethyl ether,AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 35.0°C |
Color | Yellow to Brown |
CAS Min % | 42.99, 50.22 |
CAS Max % | 49.78, 57.01 |
Physical Form | Liquid |
Quantity | 100 mL |
Linear Formula | CH3(CH2)7MgBr |
Flash Point | −35°C |
Formula Weight | 217.43 |
Concentration or Composition (by Analyte or Components) | 1.90 to 2.20M |
Show More |
Safety and Handling
Product Identifier
- Octylmagnesium bromide, 2M solution in diethyl ether
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H224-Extremely flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'