missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octyl Phenol Ethoxylate, BAKER ANALYZED™ Reagent, J.T. Baker™
93.58 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
120 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Specifications
Flash Point | 251.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 120 mL |
Specific Gravity | 1L = 1.07kg |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Octyl Phenol Ethoxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'