missing translation for 'onlineSavingsMsg'
Learn More

Octyl acetate, 99+%, Thermo Scientific Chemicals

Catalog Number p-177028
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
112-14-1
C10H20O2
172.27
MFCD00009562
YLYBTZIQSIBWLI-UHFFFAOYSA-N
n-octyl acetate, acetic acid, octyl ester, caprylyl acetate, 1-octyl acetate, acetic acid octyl ester, octyl ethanoate, n-octanyl acetate, n-octyl ethanoate, 1-octanol acetate, acetate c-8
8164
CHEBI:87495
octyl acetate
CCCCCCCCOC(C)=O
This item is not returnable. View return policy
112-14-1
C10H20O2
172.27
MFCD00009562
YLYBTZIQSIBWLI-UHFFFAOYSA-N
n-octyl acetate, acetic acid, octyl ester, caprylyl acetate, 1-octyl acetate, acetic acid octyl ester, octyl ethanoate, n-octanyl acetate, n-octyl ethanoate, 1-octanol acetate, acetate c-8
8164
CHEBI:87495
octyl acetate
CCCCCCCCOC(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 0.8680g/mL
Boiling Point 211.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3CO2(CH2)7CH3
Refractive Index 1.4175 to 1.4195
Quantity 5 g
Specific Gravity 0.868
Formula Weight 172.27
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Octyl acetate, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'