missing translation for 'onlineSavingsMsg'
Learn More

Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals

Catalog Number p-4510432
29.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5466-77-3
C18H26O3
290.40
MFCD00072582
YBGZDTIWKVFICR-UHFFFAOYNA-N
bidd:er0152, octyl methoxy cinnamate omc, unii-4y5p7mud51 component, 2s-2-ethylhexyl 2e-3-4-methoxyphenyl prop-2-enoate
11044481
[(2S)-2-ethylhexyl] (E)-3-(4-methoxyphenyl)prop-2-enoate
CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1
This item is not returnable. View return policy
5466-77-3
C18H26O3
290.40
MFCD00072582
YBGZDTIWKVFICR-UHFFFAOYNA-N
bidd:er0152, octyl methoxy cinnamate omc, unii-4y5p7mud51 component, 2s-2-ethylhexyl 2e-3-4-methoxyphenyl prop-2-enoate
11044481
[(2S)-2-ethylhexyl] (E)-3-(4-methoxyphenyl)prop-2-enoate
CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 1mg KOH/g max.
Absorbance 830 (at 307 to 308nm in methanol) min.
Color Colorless to Yellow
Density 1.0090g/mL
Flash Point 193°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5430 to 1.5470
Quantity 25 g
Merck Index 15,6859
Specific Gravity 1.009
Stabilizer 0.05 to 0.1% BHT
Formula Weight 290.39
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Octyl 4-methoxycinnamate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'