missing translation for 'onlineSavingsMsg'
Learn More

Octanophenone, 99+%, Thermo Scientific Chemicals

Catalog Number p-3779360
40.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1674-37-9
C14H20O
204.31
MFCD00003554
UDEVCZRUNOLVLU-UHFFFAOYSA-N
octanophenone, heptyl phenyl ketone, 1-octanone, 1-phenyl, caprylophenone, n-octanophenone, ketone, heptyl phenyl, 4-n-butylbutyrophenone, 1-phenyl-octan-1-one, acmc-1axlq
74291
1-phenyloctan-1-one
CCCCCCCC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1674-37-9
C14H20O
204.31
MFCD00003554
UDEVCZRUNOLVLU-UHFFFAOYSA-N
octanophenone, heptyl phenyl ketone, 1-octanone, 1-phenyl, caprylophenone, n-octanophenone, ketone, heptyl phenyl, 4-n-butylbutyrophenone, 1-phenyl-octan-1-one, acmc-1axlq
74291
1-phenyloctan-1-one
CCCCCCCC(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.93g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula C6H5CO(CH2)6CH3
Refractive Index 1.5024 to 1.5054 (20°C, 589nm)
Quantity 5 g
Formula Weight 204.31
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Octanophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'