missing translation for 'onlineSavingsMsg'
Learn More

gamma-Octanoic lactone, 98%, Thermo Scientific Chemicals

Catalog Number 10785221
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
104-50-7
C8H14O2
142.2
MFCD00005402
IPBFYZQJXZJBFQ-UHFFFAOYSA-N
gamma-octalactone, 4-octanolide, 5-butyldihydrofuran-2 3h-one, octanolide-1,4, 2 3h-furanone, 5-butyldihydro, octan-4-olide, gamma-octanolactone, 8-oxo-5-octanolide, gamma-octanoic lactone, 5-butyltetrahydro-2-furanone
7704
5-butyloxolan-2-one
CCCCC1CCC(=O)O1
This item is not returnable. View return policy
104-50-7
C8H14O2
142.2
MFCD00005402
IPBFYZQJXZJBFQ-UHFFFAOYSA-N
gamma-octalactone, 4-octanolide, 5-butyldihydrofuran-2 3h-one, octanolide-1,4, 2 3h-furanone, 5-butyldihydro, octan-4-olide, gamma-octanolactone, 8-oxo-5-octanolide, gamma-octanoic lactone, 5-butyltetrahydro-2-furanone
7704
5-butyloxolan-2-one
CCCCC1CCC(=O)O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.9800g/mL
Boiling Point 234.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4430 to 1.4450
Quantity 100 g
Specific Gravity 0.98
Formula Weight 142.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material γ-Octanoic lactone, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'