missing translation for 'onlineSavingsMsg'
Learn More

Octanal, 98%, Thermo Scientific Chemicals

Catalog Number p-7081088
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
124-13-0
C8H16O
128.22
MFCD00007029
NUJGJRNETVAIRJ-UHFFFAOYSA-N
caprylic aldehyde, caprylaldehyde, n-octanal, 1-octanal, n-octyl aldehyde, n-octaldehyde, n-caprylaldehyde, octanaldehyde, aldehyde c-8, n-octylal
454
CHEBI:17935
octanal
CCCCCCCC=O
This item is not returnable. View return policy
124-13-0
C8H16O
128.22
MFCD00007029
NUJGJRNETVAIRJ-UHFFFAOYSA-N
caprylic aldehyde, caprylaldehyde, n-octanal, 1-octanal, n-octyl aldehyde, n-octaldehyde, n-caprylaldehyde, octanaldehyde, aldehyde c-8, n-octylal
454
CHEBI:17935
octanal
CCCCCCCC=O

Octanal is an aromatic aldehyde often found in citrus oils. It is used commercially as a component in perfumes and in flavor production for the food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanal is an aromatic aldehyde often found in citrus oils. It is used commercially as a component in perfumes and in flavor production for the food industry.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. Air and moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 12°C to 15°C
Density 0.821
Boiling Point 169°C to 171°C
Flash Point 56°C (133°F)
Linear Formula CH3(CH2)6CHO
Refractive Index 1.4195
Quantity 50 g
UN Number UN1191
Sensitivity Air Sensitive
Merck Index 14,1766
Solubility Information Slightly soluble in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material Octanal
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'