missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Octahedral PTFE Stir Bar

Manufactured from PTFE encapsulated Alnico V magnet core. Fisherbrand™ Octahedral PTFE Stir Bar is autoclavable.

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.03800kg

 View more versions of this product

Product Code. 10703183

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Yes
PTFE
PTFE encapsulated Alnico V magnet core
PTFE Stirrer Bars Octahedral
75mm
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
White
Containers & Vessels / Stirrer Hot Plates
13mm
Octahedral
Videos