missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Octahedral PTFE Stir Bar

Provides stable stirring action. These PTFE magnetic stir bars are autoclavable and include a pivot ring. The flat surfaces provide a great mixing action.

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.09400kg

 View more versions of this product

Product Code. 11537782

  • £28.00 / Pack of 10


Estimated Shipment
15-06-2018
Add to basket

Description

Description

  • Stable, general-purpose magnetic stir bar
  • The flat surfaces provide a greater mixing action
  • All have pivot rings
  • AlNiCo-V magnets encased in PTFE
  • Autoclavable
Specifications

Specifications

Yes
PTFE
PTFE encapsulated Alnico V magnet core
10/Pk.
9.5mm
Octahedral
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
White
Containers & Vessels / Stirrer Hot Plates
PTFE Stirrer Bars Octahedral
38mm
Videos
Documentation