missing translation for 'onlineSavingsMsg'
Learn More

Octadecylmagnesium chloride, 0.3M solution in THF, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 436011000

Code : A0

Additional Details : CAS Number : 116980-66-6 Weight : 0.09000kg Transport : UN number : 2924 Chem class : 1b Pack group : II


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

 View more versions of this product

Product Code. 11975971

  • £53.70 / 100mL


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

Octadecylmagnesium chloride
87.82
C18H37ClMg
109-99-9
93.04
313.24

Specifications

Octadecylmagnesium chloride
87.82
0.9060g/mL
Solution, Any Turbidity Can Occur
Solubility in water: reacts.
313.24
67.0°C
100mL
109-99-9
93.04
AcroSeal™ Glass Bottle
C18H37ClMg
0.906
Solubility in water: reacts.
Yellow to brown or gray
−20°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Octadecylmagnesium chloride, 0.3M solution in THF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS