missing translation for 'onlineSavingsMsg'
Learn More

Octadecylbenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-3737098
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4445-07-2
C24H42
330.59
MFCD00048500
WSVDSBZMYJJMSB-UHFFFAOYSA-N
octadecylbenzene
CCCCCCCCCCCCCCCCCCC1=CC=CC=C1
This item is not returnable. View return policy
4445-07-2
C24H42
330.59
MFCD00048500
WSVDSBZMYJJMSB-UHFFFAOYSA-N
octadecylbenzene
CCCCCCCCCCCCCCCCCCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 32°C to 36°C
Color White to Yellow
Boiling Point 249°C (15.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5(CH2)17CH3
Quantity 1 g
Synonym 1-phenyloctadecane, benzene, octadecyl, n-octadecylbenzene, stearylbenzene, 2-phenyloctadecane, octadecane, 1-phenyl, n-octadecyl benzene, acmc-2097fm, octadecane, 1-phenyl-8ci, 1-phenyloctadecane, analytical standard
PubChem CID 78187
Formula Weight 330.59
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material Octadecylbenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'