missing translation for 'onlineSavingsMsg'
Learn More

OCT3/4, Alexa Fluor 488, clone: EM92, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-5841-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15382040

  • £93.40 / 25µg


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The EM92 monoclonal antibody reacts with mouse and human Oct3/4, encoded by the Pou5F1 gene. Oct3/4 is a POU domain-containing transcription factor that is critical for maintaining embryonic stem (ES) and induced pluripotent stem (iPS) cells in a pluripotent state, and is expressed by ES, embryonic germ cells and embryonic carcinoma cell lines. In cells of the inner cell mass (ICM), reduction of Oct3/4 expression causes dedifferentiation to trophoectoderm, whereas increased expression results in differentiation to mesoderm and primitive endoderm. Oct3/4 regulates the expression of several genes, including FGF-4, UTF1, Sox2, Fbx15, Rex1 and osteopontin through distinct mechanisms. Furthermore, Oct3/4 frequently acts synergistically with Sox2 to regulate target gene expression, as is the case with FGF-4. It has been demonstrated that Oct3/4 expression in ES cells can be negatively regulated by either treatment with retinoic acid, or by removal of leukemia-inhibitory factor (LIF).Applications Reported: This EM92 antibody has been reported for use in intracellular staining followed by flow cytometric analysis and immunocytochemistry.Applications Tested: This EM92 antibody has been tested by microscopy or intracellular staining and flow cytometric analysis of F9 cells using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Please see Best Protocols for Staining Protocol (refer to Protocol B: One step protocol for intracellular (nuclear) proteins). This antibody can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This antibody has also been tested by immunocytochemistry on formaldehyde fixed and permeabilized cells at less than or equal to 10μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

OCT3/4
EM92
Alexa Fluor 488
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Immunocytochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
OCT3, OCT4, octamer-binding transcription factor, POU5F1
IgG2a
RUO
Primary
Murine
Videos