missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse OCIL/CLEC2d Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | OCIL/CLEC2d |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
OCIL/CLEC2d Polyclonal specifically detects OCIL/CLEC2d in Mouse samples. It is validated for Western Blot.Specifications
OCIL/CLEC2d | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CLAXLLT-1, CLEC2D, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1, LLT1Osteoclast inhibitory lectin, OCILmember D | |
CLEC2D | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q91V08 | |
29121 | |
E. coli-derived recombinant mouse OCIL/CLEC2d Ala65-Ser207 Accession # Q91V08 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse OCIL/CLEC2d in direct ELISAs and Western blots. In Western blots, approximately 35% cross-reactivity with recombinant mouse OCILRP2 is observed and less than 2% cross-reactivity with recombinant human OCIL is observed. |
For Research Use Only