missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ocean Blue, SE
GREENER_CHOICE

Product Code. 18767496
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 18767496

Brand: Tocris Bioscience™ 6489/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Blue fluorescent dye; supplied as NHS ester

Ocean Blue, SE is a blue fluorescent dye. Conjugates of this dye are strongly fluorescent even at neutral pH. Excitation maximum = 405 nm; emission maximum = 455 nm; extinction coefficient = 36000 M-1cm-1; correction factor A280 = 0.15.

Please see the protocol for further information on protein/antibody labeling and conjugation.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Carboxy-6,8-difluoro-7-hydroxycoumaryl succinimidyl ester
CAS 215868-33-0
Quantity 5 mg
Target Standard Fluorescein, Coumarin and Rhodamine-based Dyes
Molecular Formula C14H7F2NO7
Formula Weight 339.2
Product Type Blue Fluorescent Dye
Purity 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.