missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse OCAM/NCAM2 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | OCAM/NCAM2 |
---|---|
Clone | 125101 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
OCAM/NCAM2 Monoclonal specifically detects OCAM/NCAM2 in Mouse samples. It is validated for Western Blot.Specifications
OCAM/NCAM2 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
NCAM2, N-CAM-2, NCAM-2, NCAM21MGC51008, neural cell adhesion molecule 2, neural cell adhesion molecule 2 precursor10, Rb-8, RNCAM | |
NCAM2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
125101 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
O35136 | |
4685 | |
Mouse myeloma cell line NS0-derived recombinant mouse OCAM/NCAM2 Leu20-Gly700 (predicted) Accession # O35136 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse OCAM/NCAM2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhMCAM, recombinant mouse (rm) NCAM-L1, or rmMAdCAM-1 is observed. |
For Research Use Only