Learn More
Leica Microsystems Objective, HI-Plan
Economical, high quality microscope objective designed for visual fields up to 20mm. Leica Microsystems™ Objective HI-Plan features good chromatic correction at two wavelengths, flatness over the whole field of view and sharp image edges.
Brand:  Leica Microsystems 11506240
Code : 03
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11974419
  • £1113.81 / Each
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Description
Description
The Leica DM1000 is the ideal instrument for biological, medical and clinical laboratories. The instrument can be configured to meet specific requirements by choosing from a selection of observation tubes, ergonomic modules, objectives and adjustable controls. Please note a complete system comprises the main microscope unit, an observation tube and either the brightfield or phase contrast outfit. Fluorescence outfits also require the brightfield or phase contrast outfit to be ordered.
Specifications
Specifications
Objective HI-Plan
40X
1 Ea.