Learn More
Carl Zeiss™ Objective A Achromat
Use these universal objectives for routine research. Zeiss™ A-Plan Objectives provide rich contrast and can be used with eyepieces with a field of view up to 23mm.
Brand: Carl Zeiss™ 421091-9912-000
Code : NEW
Additional Details : CAS Number : 195371-10-9 Weight : 0.64980kg
Description
- This class of objectives provides an attractive alternative to Achroplan objectives, both in routine and research applications
- Allow contrasting methods ranging from brightfield to phase contrast and for excitation wavelength within the visible spectrum
- Can be used for epifluorescence work
Specifications
Oil PH3 M27 | |
Numerical Aperture: 1.25mm; Oil PH3 M27 | |
100X | |
1 Ea. |