Learn More
OBAA, Tocris Bioscience™
Phospholipase A2 inhibitor
Brand:  Tocris (Bio-Techne) 060650
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13487803
  • £461.52 / 50mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
A potent inhibitor of phospholipase A2 (IC50 = 70 nM). Reduces bronchospasm in guinea pigs in vivo.
Specifications
Chemical Identifiers
221632-26-4
428.657
4-(4-Octadecylphenyl)-4-oxobutenoic acid
C28H44O3
ZBESASFHIWDSCJ-WCWDXBQESA-N
5353842
Specifications
OBAA
>98%
4-(4-Octadecylphenyl)-4-oxobutenoic acid
CCCCCCCCCCCCCCCCCCC1=CC=C(C=C1)C(=O)C=CC(=O)O
428.657
428.65
221632-26-4
C28H44O3
ZBESASFHIWDSCJ-WCWDXBQESA-N
(E)-4-(4-octadecylphenyl)-4-oxobut-2-enoic acid
5353842