Learn More
O-Xylene, Pure, Fisher Chemical™
Brand: Fisher Chemical X/0300/08
Code : 21
Additional Details : CAS Number : 95-47-6 Weight : 0.43000kg Transport : UN number : 1307 Chem class : 3 Pack group : III
Quantity | 500mL |
---|
Chemical Identifiers
95-47-6 | |
106.168 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
7237 | |
1,2-xylene |
C8H10 | |
8519 | |
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
CHEBI:28063 | |
CC1=CC=CC=C1C |
Specifications
95-47-6 | |
8519 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
1,2-xylene | |
7237 | |
106.17g/mol | |
Amber glass bottle | |
143°C | |
-25°C |
C8H10 | |
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
CC1=CC=CC=C1C | |
106.168 | |
CHEBI:28063 | |
Liquid | |
3.7 | |
Colorless | |
500mL |
Safety and Handling
- o-Xylene
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.