missing translation for 'onlineSavingsMsg'
Learn More

O-(Trimethylsilyl)hydroxylamine, 97%, ACROS Organics™

Brand:  Acros Organics 367950010

Code : 29

Additional Details : CAS Number : 22737-36-6 Weight : 0.05100kg Transport : UN number : 1993 Chem class : 1 Pack group : II


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10541882

  • £37.90 / 1g


Estimated Shipment
11-05-2018
Add to basket


Specifications

Chemical Identifiers

O-(Trimethylsilyl)hydroxylamine
22737-36-6
100.0
97%
105.21
Aminoxytrimethylsilane
97%
96.0
≥96.0%
C3H11NOSi
MFCD00010872

Specifications

O-(Trimethylsilyl)hydroxylamine
Clear colorless
97%
96.0
Authentic
1.4040 to 1.4060 (20°C, 589nm)
≥96.0%
C3H11NOSi
MFCD00010872
0.86g/mL
1g
22737-36-6
100.0
Glass bottle
Liquid
97%
105.21
Aminoxytrimethylsilane
Videos
SDS