missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4089550
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
932-31-0, 60-29-7
C7H7BrMg
195.34
MFCD00010350
DVXDIGKNJYSMFM-UHFFFAOYSA-M
o-tolylmagnesium bromide, bromo-o-tolylmagnesium, bromo 2-methylphenyl magnesium, 2-tolylmagnesium bromide, o-methylphenylmagnesium bromide, 2-methylphenylmagnesium bromide, magnesium,bromo 2-methylphenyl, o-tolyl magnesium bromide, tolylmagnesium bromide, o-tolylmagnesiumbromid, grignard reagent
101919
magnesium;methylbenzene;bromide
CC1=CC=CC=C1[Mg]Br
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
932-31-0, 60-29-7
C7H7BrMg
195.34
MFCD00010350
DVXDIGKNJYSMFM-UHFFFAOYSA-M
o-tolylmagnesium bromide, bromo-o-tolylmagnesium, bromo 2-methylphenyl magnesium, 2-tolylmagnesium bromide, o-methylphenylmagnesium bromide, 2-methylphenylmagnesium bromide, magnesium,bromo 2-methylphenyl, o-tolyl magnesium bromide, tolylmagnesium bromide, o-tolylmagnesiumbromid, grignard reagent
101919
magnesium;methylbenzene;bromide
CC1=CC=CC=C1[Mg]Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown to Gray
Density 1.0130g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3C6H4MgBr
Quantity 100 mL
Specific Gravity 1.013
Formula Weight 195.34
Physical Form Solution
Chemical Name or Material o-Tolylmagnesium bromide, 2M solution in diethyl ether
compliance-icons compliance-icons compliance-icons
Product Identifier
  • o-Tolylmagnesium bromide, 2M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'