missing translation for 'onlineSavingsMsg'
Learn More

o-Toluoyl chloride, 99%, Thermo Scientific Chemicals

Catalog Number p-201130
13.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
933-88-0
C8H7ClO
154.59
MFCD00000668
GPZXFICWCMCQPF-UHFFFAOYSA-N
o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride
70276
2-methylbenzoyl chloride
CC1=CC=CC=C1C(Cl)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
933-88-0
C8H7ClO
154.59
MFCD00000668
GPZXFICWCMCQPF-UHFFFAOYSA-N
o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride
70276
2-methylbenzoyl chloride
CC1=CC=CC=C1C(Cl)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Pink to Yellow
Density 1.1800g/mL
Boiling Point 88°C to 90°C (12.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4COCl
Refractive Index 1.5540 to 1.556
Quantity 5 g
Beilstein 09, 464
Specific Gravity 1.18
Solubility Information Solubility in water: decomposes
Formula Weight 154.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material o-Toluoyl chloride, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'