missing translation for 'onlineSavingsMsg'
Learn More

o-Toluidine, 99%, Thermo Scientific Chemicals

Catalog Number p-7021318
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
95-53-4
C7H9N
107.156
MFCD00007730
RNVCVTLRINQCPJ-UHFFFAOYSA-N
o-toluidine, 2-toluidine, o-tolylamine, 2-aminotoluene, 2-methylbenzenamine, ortho-toluidine, o-methylaniline, benzenamine, 2-methyl, o-aminotoluene, o-methylbenzenamine
7242
CHEBI:66892
2-methylaniline
CC1=CC=CC=C1N
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
95-53-4
C7H9N
107.156
MFCD00007730
RNVCVTLRINQCPJ-UHFFFAOYSA-N
o-toluidine, 2-toluidine, o-tolylamine, 2-aminotoluene, 2-methylbenzenamine, ortho-toluidine, o-methylaniline, benzenamine, 2-methyl, o-aminotoluene, o-methylbenzenamine
7242
CHEBI:66892
2-methylaniline
CC1=CC=CC=C1N

o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.

Solubility
Soluble in water(15g/L).

Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point -24°C
Density 1.004
Boiling Point 199°C to 200°C
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.572
Quantity 250 g
UN Number UN1708
Beilstein 741981
Sensitivity Air and light sensitive
Merck Index 14,9536
Solubility Information Soluble in water(15g/L).
Formula Weight 107.16
Percent Purity 99%
Chemical Name or Material o-Toluidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'