missing translation for 'onlineSavingsMsg'
Learn More

o-Tolualdehyde, 98%, stabilized, Thermo Scientific Chemicals

Catalog Number p-201066
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
529-20-4, 123-31-9
C8H8O
120.15
MFCD00003338
BTFQKIATRPGRBS-UHFFFAOYSA-N
o-tolualdehyde, 2-tolualdehyde, 2-formyltoluene, o-toluylaldehyde, o-toluic aldehyde, o-tolylaldehyde, o-methylbenzaldehyde, benzaldehyde, 2-methyl, 2-methyl-benzaldehyde, o-methylbenazldehyde
10722
CHEBI:27434
2-methylbenzaldehyde
CC1=CC=CC=C1C=O
This item is not returnable. View return policy
529-20-4, 123-31-9
C8H8O
120.15
MFCD00003338
BTFQKIATRPGRBS-UHFFFAOYSA-N
o-tolualdehyde, 2-tolualdehyde, 2-formyltoluene, o-toluylaldehyde, o-toluic aldehyde, o-tolylaldehyde, o-methylbenzaldehyde, benzaldehyde, 2-methyl, 2-methyl-benzaldehyde, o-methylbenazldehyde
10722
CHEBI:27434
2-methylbenzaldehyde
CC1=CC=CC=C1C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -35°C
Color Colorless to Yellow
Density 1.0390g/mL
Boiling Point 199°C to 200°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CHO
Refractive Index 1.5460 to 1.548
Quantity 25 g
Beilstein 07, 295
Merck Index 15, 9686
Specific Gravity 1.039
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol and ether
Stabilizer 0.1% hydroquinone
Formula Weight 120.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material o-Tolualdehyde, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • o-Tolualdehyde, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'