missing translation for 'onlineSavingsMsg'
Learn More

o-Thiocresol, 98%, Thermo Scientific Chemicals

Catalog Number p-7026458
18.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
137-06-4
C7H8S
124.20
MFCD00004838
LXUNZSDDXMPKLP-UHFFFAOYSA-N
o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl
8712
2-methylbenzenethiol
CC1=CC=CC=C1S
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
137-06-4
C7H8S
124.20
MFCD00004838
LXUNZSDDXMPKLP-UHFFFAOYSA-N
o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl
8712
2-methylbenzenethiol
CC1=CC=CC=C1S

o-Thiocresol is used as a thiol for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Thiocresol is used as a thiol for proteomics research.

Solubility
Soluble in ether and ethanol. Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 10°C to 12°C
Density 1.054
Boiling Point 195°C
Flash Point 63°C (145°F)
Refractive Index 1.58
Quantity 5 g
UN Number UN2810
Beilstein 774068
Sensitivity Air Sensitive
Merck Index 14,9325
Solubility Information Soluble in ether and ethanol. Insoluble in water.
Formula Weight 124.21
Percent Purity 98%
Chemical Name or Material o-Thiocresol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'