missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-L-serine, 97%, Thermo Scientific Chemicals

Catalog Number 10814341
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
18822-58-7
DDCPKNYKNWXULB-YFKPBYRVSA-N
h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh
7004940
(2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid
CC(C)(C)OCC(C(=O)O)N
This item is not returnable. View return policy
18822-58-7
DDCPKNYKNWXULB-YFKPBYRVSA-N
h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh
7004940
(2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid
CC(C)(C)OCC(C(=O)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 225.2°C to 228.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Molecular Formula C7H15NO3
Quantity 5 g
Specific Rotation − 15.50
Molecular Weight (g/mol) 161.2
Formula Weight 161.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material O-tert-Butyl-L-serine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'