missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ O-tert-Butyl-D-tyrosine, 95%

Brand:  Alfa Aesar™ H63130.06

Code : NEW

Additional Details : CAS Number : 186698-58-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15479648

  • £267.12 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

O-tert-Butyl-D-tyrosine
237.29
H-D-Tyr(tBu)-OH; (R)-2-Amino-3-(4-tert-butoxyphenyl)propionic acid
186698-58-8
MFCD00077112

Specifications

O-tert-Butyl-D-tyrosine
Solid
237.29
H-D-Tyr(tBu)-OH; (R)-2-Amino-3-(4-tert-butoxyphenyl)propionic acid
186698-58-8
95%
MFCD00077112
5g
Videos
SDS