missing translation for 'onlineSavingsMsg'
Learn More

o-Phenetidine, 99%, Thermo Scientific Chemicals

Catalog Number p-181150
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
94-70-2
C8H11NO
137.18
MFCD00007689
ULHFFAFDSSHFDA-UHFFFAOYSA-N
o-phenetidine, 2-aminophenetole, benzenamine, 2-ethoxy, o-ethoxyaniline, 2-ethoxybenzenamine, o-aminophenetole, 2-ethoxyphenylamine, ethoxyaniline, 2-phenetidine, benzenamine, ar-ethoxy
7203
2-ethoxyaniline
CCOC1=CC=CC=C1N
This item is not returnable. View return policy
94-70-2
C8H11NO
137.18
MFCD00007689
ULHFFAFDSSHFDA-UHFFFAOYSA-N
o-phenetidine, 2-aminophenetole, benzenamine, 2-ethoxy, o-ethoxyaniline, 2-ethoxybenzenamine, o-aminophenetole, 2-ethoxyphenylamine, ethoxyaniline, 2-phenetidine, benzenamine, ar-ethoxy
7203
2-ethoxyaniline
CCOC1=CC=CC=C1N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -20.0°C
Color Red
Density 1.0500g/mL
Boiling Point 230.0°C to 233.0°C
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OC6H4NH2
Refractive Index 1.5540 to 1.5560
Quantity 5 mL
Merck Index 15, 7341
Specific Gravity 1.05
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 137.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material o-Phenetidine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • o-Phenetidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'