missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Methylisourea hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.
Solubility
Soluble in organic solvents, water, methanol and ethanol.
Notes
Incompatible with strong oxidizing agents.
O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.
Solubility
Soluble in organic solvents, water, methanol and ethanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∼116°C (decomposition) |
Quantity | 10 g |
Beilstein | 3909619 |
Solubility Information | Soluble in organic solvents,water,methanol and ethanol. |
Formula Weight | 110.54 |
Percent Purity | 98% |
Chemical Name or Material | O-Methylisourea hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'