missing translation for 'onlineSavingsMsg'
Learn More

o-Cresol, ≥99, Reagent Grade, Honeywell™

Catalog Number p-7120635
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
95-48-7
C7H8O
108.14
MFCD00002226
QWVGKYWNOKOFNN-UHFFFAOYSA-N
o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
335
CHEBI:28054
2-methylphenol
CC1=CC=CC=C1O
This item is not returnable. View return policy
95-48-7
C7H8O
108.14
MFCD00002226
QWVGKYWNOKOFNN-UHFFFAOYSA-N
o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
335
CHEBI:28054
2-methylphenol
CC1=CC=CC=C1O
Melting Point 30.99°C (1013hPa)
Color Colorless
Density 1.05g/cu.cm at 20°C
Boiling Point 191°C (1013 hPa)
Flash Point 81°C (177.8°F)
Ignition Point 1110°F
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3C6H4OH
Quantity 100 g
UN Number UN3455
Beilstein 506917
Solubility Information 26g/L at 25°C Solubility in water
Vapor Density 3.72 (vs air)
Formula Weight 108.14g/mol
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material o-Cresol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'