missing translation for 'onlineSavingsMsg'
Learn More

O-bromophenoxyacetic acid, 98%, ACROS Organics™

Brand:  Acros Organics 379210250

Code : 29

Additional Details : CAS Number : 1879-56-7 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10568463

  • £52.20 / 25g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

2-Bromophenoxyacetic acid
97.5
≥97.5%
C8H7BrO3
231.05
1879-56-7
100.0
98%
BrC6H4OCH2CO2H
MFCD00234407

Specifications

2-Bromophenoxyacetic acid
97.5
Authentic
141° to 145°C
White to light pink
≥97.5%
C8H7BrO3
231.05
1879-56-7
100.0
Glass bottle
25g
Crystalline Powder
98%
BrC6H4OCH2CO2H
MFCD00234407
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bromophenoxyacetic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS