missing translation for 'onlineSavingsMsg'
Learn More

O-Benzylhydroxylamine, 97%, Thermo Scientific Chemicals

Catalog Number 10795983
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
622-33-3
C7H9NO
123.15
MFCD00221709
XYEOALKITRFCJJ-UHFFFAOYSA-N
benzyloxyamine, hydroxylamine, o-phenylmethyl, hydroxylamine, o-benzyl, o-benzyl-hydroxylamine, benzyl-o-hydroxylamine, oxybenzylamine, o-phenylmethyl hydroxylamine, o-benzyloxylamine, phenylmethoxyamine, benzyloxy amine
102313
O-benzylhydroxylamine
C1=CC=C(C=C1)CON
This item is not returnable. View return policy
622-33-3
C7H9NO
123.15
MFCD00221709
XYEOALKITRFCJJ-UHFFFAOYSA-N
benzyloxyamine, hydroxylamine, o-phenylmethyl, hydroxylamine, o-benzyl, o-benzyl-hydroxylamine, benzyl-o-hydroxylamine, oxybenzylamine, o-phenylmethyl hydroxylamine, o-benzyloxylamine, phenylmethoxyamine, benzyloxy amine
102313
O-benzylhydroxylamine
C1=CC=C(C=C1)CON

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.1900g/mL
Boiling Point 110°C to 112°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2ONH2
Refractive Index 1.5390 to 1.5410 (20°C, 589nm)
Quantity 5 g
Beilstein 06, IV, 2562
Specific Gravity 1.19
Solubility Information Solubility in water: soluble
Formula Weight 123.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material O-Benzylhydroxylamine
Show More Show Less
compliance-icons
Product Identifier
  • O-Benzylhydroxylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'