missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl-L-serine, 99%, ACROS Organics™

Brand:  Acros Organics 440930050

Code : 29

Additional Details : CAS Number : 4726-96-9 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 11995511

  • £76.90 / 5g


Estimated Shipment
18-06-2018
Add to basket

Specifications

Chemical Identifiers

O-Benzyl-L-serine
98.5
≥98.5%
C10H13NO3
H-Ser(Bzl)-OH
4726-96-9
100.0
99%
195.22

Specifications

O-Benzyl-L-serine
98.5
Authentic
Powder
≥98.5%
1% Max.
195.22
5g
4726-96-9
100.0
Glass bottle
+20.0° to +25.0° (20°C, 589nm) (c=1 in 8:2 AcOH/1M HCl)
99%
C10H13NO3
White to off-white
H-Ser(Bzl)-OH
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • O-Benzyl-L-serine

Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.

SDS