missing translation for 'onlineSavingsMsg'
Learn More

o-Anisidine, 99+%, Thermo Scientific Chemicals

Catalog Number p-60955
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
90-04-0
C7H9NO
123.15
MFCD00007688
VMPITZXILSNTON-UHFFFAOYSA-N
o-anisidine, 2-anisidine, ortho-anisidine, 2-aminoanisole, o-methoxyaniline, o-aminoanisole, benzenamine, 2-methoxy, 2-methoxybenzenamine, o-methoxyphenylamine, o-anisylamine
7000
CHEBI:82288
2-methoxyaniline
COC1=CC=CC=C1N
This item is not returnable. View return policy
90-04-0
C7H9NO
123.15
MFCD00007688
VMPITZXILSNTON-UHFFFAOYSA-N
o-anisidine, 2-anisidine, ortho-anisidine, 2-aminoanisole, o-methoxyaniline, o-aminoanisole, benzenamine, 2-methoxy, 2-methoxybenzenamine, o-methoxyphenylamine, o-anisylamine
7000
CHEBI:82288
2-methoxyaniline
COC1=CC=CC=C1N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 3.0°C to 6.0°C
Density 1.0920g/mL
Boiling Point 225.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4NH2
Refractive Index 1.5730 to 1.5750
Quantity 5 mL
Beilstein 13, 358
Merck Index 14, 667
Specific Gravity 1.092
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in dilute mineral acid, miscible with alcohol and ether, soluble in acetone and benzene
Formula Weight 123.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material o-Anisidine, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • o-Anisidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'