missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Anisidine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.
Solubility
Soluble in dilute inorganic acid, alcohol, diethyl ether. Not miscible in water.
Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents, acid anhydrides, chloroformates, acids.
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.
Solubility
Soluble in dilute inorganic acid, alcohol, diethyl ether. Not miscible in water.
Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents, acid anhydrides, chloroformates, acids.
Specifications
Melting Point | 5°C to 6°C |
Density | 1.092 g/mL |
Boiling Point | 224°C to 225°C |
Flash Point | 98°C (208°F) |
Assay Percent Range | 99% |
Refractive Index | 1.574 |
Quantity | 250 g |
UN Number | UN2431 |
Beilstein | 386210 |
Sensitivity | Light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'