missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-(4-Methoxybenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Color | White |
Molecular Formula | C8H11NO2 |
Quantity | 1 g |
InChI Key | MVSMBIBGGPSEHQ-UHFFFAOYSA-N |
SMILES | COC1=CC=C(C=C1)CON |
Molecular Weight (g/mol) | 153.181 |
Formula Weight | 153.18 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | O-(4-Methoxybenzyl)hydroxylamine hydrochloride |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'