missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nylon sheet, 12.7mm (0.5 in.) thick

Brand:  Alfa Aesar™ 045218.HB

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300x300mm

Product Code. 15492207

  • £67.95 / Each


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Specifications

Nylon sheet
12.7mm (0.5 in.) thick
MFCD00133998
300x300mm
Videos
SDS