missing translation for 'onlineSavingsMsg'
Learn More

Millipore™ Nylon Plain Surface Net Filter

Brand:  Millipore™ NY8H00010

Code : 71

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10517302

  • £311.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

White
NY8H
Net Filter
1Pack
For use in Collection of algae & cells, particle analysis, large particulate filtration, prefiltration of solvents, paint monitoring
180μm
3m x 30cm
Plain
14%
100°C
Nylon
Net Filter
Videos