missing translation for 'onlineSavingsMsg'
Learn More

Millipore™ Nylon Plain Surface Net Filter

Brand:  Millipore™ NY6009000

Code : 71

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10317663

  • £194.00 / Pack of 50


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

White
NY60
Net Filter
50Pack
For use in Collection of algae & cells, particle analysis, large particulate filtration, prefiltration of solvents, paint monitoring
60μm
90mm
Plain
41%
100°C
Nylon
Net Filter
Videos