Learn More
Masterflex™ Nylon Female Luer Fitting
Use these low-profile hose barbs with semi-rigid tubing such as nylon, polyethylene and polypropylene. Nylon Female Luer Fittings feature a smooth bore hose barb to reduce flow turbulence and risk of line contamination.
Brand: Masterflex™ 45502-05
Code : NEW
Additional Details : Weight : 0.00940kg
Description
- Standard hose barb for use with flexible tubing
- Smooth bore hose barb to reduce flow turbulence and risk of line contamination
- Wide-bore hose barb for enhanced fluid path flow
Specifications
Female Luer Fitting | |
Nylon | |
Fits Female luer to 1/8 in. wide-bore hose barb | |
25 Pack |