missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Filter Storage Bottles

Save time and minimize contamination with these radiation-sterilized filter storage bottles. Excellent as single-use storage containers.

Brand:  Thermo Scientific™ 455-0250

Code : 55

Additional Details : Weight : 2.40000kg

 View more versions of this product

Product Code. 10636591

  • £113.00 / Pack of 24


In Stock 7
Add to basket

Disclaimers:

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.


Description

Description

  • Headspace allows for extra serum or other additives to be added to the filtrate
  • Leakproof† screw caps to eliminate pH shift in bottle for longer life of stored media
  • Sterilized by gamma radiation and individually bagged for a 5-year sterile shelf life
  • Non-pyrogenic and non-cytotoxic
Specifications

Specifications

250mL
24
Wide
Wide
24
Bottle: virgin clear polystyrene; Closure: white pigmented virgin high density polyethylene
Sterile
Videos
Documentation