missing translation for 'onlineSavingsMsg'
Learn More

NVP DPP 728 dihydrochloride, Tocris Bioscience™

Potent, orally active dipeptidyl peptidase IV (DPP-IV) inhibitor

Brand:  Tocris (Bio-Techne) 350610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13463094

  • £188.76 / 10mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Potent, orally active dipeptidyl peptidase (DPP)-IV inhibitor (Ki = 11 nM, IC50 = 14 nM) that displays > 15 000-fold selectivity over DPP-II and a range of proline-cleaving proteases. Exhibits antidiabetic activity in vivo; improves glucose tolerance, increases glucagon-like peptide 1 (GLP-1) and insulin levels, augments insulin secretion and GLUT2 levels, and reduces (preserves) islet size.
Specifications

Chemical Identifiers

207556-62-5
371.266
pwktkamjur, unii-pwktkamjur, nvp-dpp-728 dihydrochloride, nvp dpp 728 dihydrochloride, dpp-728, nvp-dpp-728, 6-2-2-2s-2-cyano-1-pyrrolidinyl-2-oxoethyl amino eth, s-6-2-2-2-cyanopyrrolidin-1-yl-2-oxoethyl amino ethyl amino nicotinonitrile dihydrochloride, 1-2-5-cyano-2-pyridinyl amino ethylamino acetyl-2-pyrrolidinecarbonitrile dihydrochloride, 2s, 2-pyrrolidinecarbonitrile, 1-2-5-cyano-2-pyridinyl amino ethyl amino acetyl-, dihydrochloride, 2s
6-[2-[[2-[(2S)-2-cyanopyrrolidin-1-yl]-2-oxoethyl]amino]ethylamino]pyridine-3-carbonitrile;dihydrochloride
C15H20Cl2N6O
VNACOBVZDCLAEV-GXKRWWSZSA-N
9799555
C1CC(N(C1)C(=O)CNCCNC2=NC=C(C=C2)C#N)C#N.Cl.Cl

Specifications

NVP DPP 728 dihydrochloride
207556-62-5
pwktkamjur, unii-pwktkamjur, nvp-dpp-728 dihydrochloride, nvp dpp 728 dihydrochloride, dpp-728, nvp-dpp-728, 6-2-2-2s-2-cyano-1-pyrrolidinyl-2-oxoethyl amino eth, s-6-2-2-2-cyanopyrrolidin-1-yl-2-oxoethyl amino ethyl amino nicotinonitrile dihydrochloride, 1-2-5-cyano-2-pyridinyl amino ethylamino acetyl-2-pyrrolidinecarbonitrile dihydrochloride, 2s, 2-pyrrolidinecarbonitrile, 1-2-5-cyano-2-pyridinyl amino ethyl amino acetyl-, dihydrochloride, 2s
C1CC(N(C1)C(=O)CNCCNC2=NC=C(C=C2)C#N)C#N.Cl.Cl
371.266
371.27
10mg
C15H20Cl2N6O
VNACOBVZDCLAEV-GXKRWWSZSA-N
6-[2-[[2-[(2S)-2-cyanopyrrolidin-1-yl]-2-oxoethyl]amino]ethylamino]pyridine-3-carbonitrile;dihydrochloride
9799555
>98%
Videos