Nur77 Mouse anti-Mouse, PerCP-eFluor 710, Clone: 12.14, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 46-5965-80

25UG Anti-Mouse Nur77 PerCP-eFluor 710

Code: NEW

Additional Details:
Additional Details: Weight: 0.09500kg



Disclaimers: For Research Use Only.

Product Code. 15509326

Quantity Price
1 £ 84.99 / 25µg
Estimated Shipment date
from Supplier 28-03-2017
Add to basket


Description and Specification

Specification

Antigen Nur77
Format Conjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gene Alias NGFI-B, NR4A1, TR3, NAK1
Host Species Mouse
Applications Flow Cytometry (Intracellular Staining)
Isotype IgG1
Quantity 25μg
Regulatory Status RUO
Species Reactivity Mouse
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 12.14
Concentration 0.2mg/mL
Conjugate PerCP-eFluor 710

This 12.14 monoclonal antibody reacts with mouse Nur77 (also known as NR4A1, TR3, NGFI-B, or NAK1), an inducible orphan nuclear receptor. Expressed in thymocytes and T cell lines, Nur77 promotes apoptosis and plays a role in thymocyte negative selection. Additionally, Nur77 has been shown to be critical for steroid biosynthesis in Leydig cells as well as for the effects of dopamine. In addition, Nur77 has been shown to interact with FoxP3 in regulatory T cells. However, our results with this antibody do not correlate with this observation.